The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008325.4 | NP_032351.2 | 6 - 7 | 1009 | 72 |
NM_172997.1 | NP_766585.1 | 5 - 6 | 868 | 72 | |
GenBank mRNA | AK031414.1 | - | 5 - 6 | 663 | 72 |
AK045330.1 | - | 4 - 5 | 496 | 72 | |
AK155627.1 | - | 6 - 7 | 1010 | 72 | |
AK169998.1 | - | 5 - 6 | 667 | 72 | |
BC057935.1 | - | 5 - 6 | 630 | 72 | |
L34111.1 | - | 5 - 6 | 583 | 72 |