Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmas
Entrez Gene ID
12764
Gene Name
cytidine monophospho-N-acetylneuraminic acid synthetase
Gene Aliases
AW208911, CMPNeu5Ac, D6Bwg0250e
Chromosome Location
Chr.6: 142756686 - 142775714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009908.2 NP_034038.2 7 - 8 1198 68
GenBank mRNA AJ006215.1 - 7 - 8 1111 68
AK087150.1 - 4 - 5 500 68
AK155730.1 - 7 - 8 1151 68

View Full Product Details