Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif3f
Entrez Gene ID
66085
Gene Name
eukaryotic translation initiation factor 3, subunit F
Gene Aliases
0610037M02Rik, AA409853, Eif3s5
Chromosome Location
Chr.7: 108934415 - 108941942 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025344.2 NP_079620.2 6 - 7 911 92
GenBank mRNA AK002778.1 - 6 - 7 910 92
AK003372.1 - 6 - 7 909 92
AK010333.1 - 6 - 7 904 92
AK088537.1 - 6 - 7 911 92
AK136797.1 - - 910 92
AK152037.1 - 6 - 7 912 92
AY061808.1 - 3 - 4 255 92
BC070473.1 - 6 - 7 893 92
BC083190.1 - 6 - 7 896 92

View Full Product Details