Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx56
Entrez Gene ID
52513
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 56
Gene Aliases
2600001H07Rik, D11Ertd619e, Noh61
Chromosome Location
Chr.11: 6257545 - 6267729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026538.3 NP_080814.1 4 - 5 563 102
GenBank mRNA AK011136.1 - 4 - 5 573 102
AK076047.1 - 4 - 5 581 102
AK076975.1 - 4 - 5 563 102
AK077514.1 - 4 - 5 576 102
AK145628.1 - 4 - 5 604 102
AK159793.1 - 4 - 5 567 102
AK169654.1 - 4 - 5 578 102
BC018291.1 - 4 - 5 557 102

View Full Product Details