Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk5rap3
Entrez Gene ID
80280
Gene Name
CDK5 regulatory subunit associated protein 3
Gene Aliases
1810007E24Rik, BC002318, C53, C81486, HSF-27, IC53, LZAP, MST016
Chromosome Location
Chr.11: 96907763 - 96916530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK050014.1 - 13 - 14 1555 75
AK075771.1 - 13 - 14 1542 75
AK075791.1 - 13 - 14 1541 75
AK149765.1 - 13 - 14 1569 75
AK153951.1 - 13 - 14 1460 75
AK161834.1 - 11 - 12 1400 75
AK165494.1 - 12 - 13 1492 75
AK166476.1 - 13 - 14 1569 75
AK189168.1 - 2 - 3 288 75
BC002318.1 - 13 - 14 1539 75

View Full Product Details