The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133239.2 | NP_573502.2 | 1 - 2 | 229 | 77 |
GenBank mRNA | AF406641.1 | - | 1 - 2 | 194 | 77 |
AF492496.1 | - | 1 - 2 | 253 | 77 | |
AY450552.1 | - | 1 - 2 | 194 | 77 |