The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144815.2 | NP_659064.1 | 3 - 4 | 451 | 62 |
GenBank mRNA | AK140387.1 | - | 3 - 4 | 472 | 62 |
BC014705.1 | - | 3 - 4 | 460 | 62 |