Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cecr5
Entrez Gene ID
214932
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
A930002G03Rik
Chromosome Location
Chr.6: 120509494 - 120531364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144815.2 NP_659064.1 6 - 7 749 60
GenBank mRNA AK140387.1 - 6 - 7 770 60
BC014705.1 - 6 - 7 758 60

View Full Product Details