The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172205.1 | NP_001165676.1 | 4 - 5 | 395 | 82 |
NM_001172206.1 | NP_001165677.1 | 4 - 5 | 392 | 82 | |
NM_001290726.1 | NP_001277655.1 | 5 - 6 | 490 | 82 | |
NM_001290727.1 | NP_001277656.1 | 3 - 4 | 271 | 82 | |
NR_033310.1 | 3 - 4 | 274 | 82 | ||
GenBank mRNA | AK149839.1 | - | 4 - 5 | 375 | 82 |
AK155555.1 | - | 3 - 4 | 251 | 82 | |
AK156376.1 | - | 4 - 5 | 375 | 82 | |
AK156622.1 | - | 3 - 4 | 657 | 82 | |
AK157607.1 | - | 4 - 5 | 375 | 82 | |
BC027152.1 | - | 3 - 4 | 238 | 82 | |
CX209955.1 | - | 3 - 4 | 254 | 82 |