The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146065.1 | NP_666177.1 | 10 - 11 | 1205 | 83 |
GenBank mRNA | AK039648.1 | - | - | 1197 | 83 |
AK047901.1 | - | - | 1181 | 83 | |
AK154588.1 | - | - | 1196 | 83 | |
AK155046.1 | - | 10 - 11 | 1196 | 83 | |
AK157628.1 | - | - | 1235 | 83 | |
BC026483.1 | - | 10 - 11 | 1205 | 83 |