Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a25
Entrez Gene ID
227731
Gene Name
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
Gene Aliases
1110030N17Rik, MCSC, mKIAA1896
Chromosome Location
Chr.2: 32414487 - 32451470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164357.1 NP_001157829.1 3 - 4 617 84
NM_001290558.1 NP_001277487.1 3 - 4 486 84
GenBank mRNA AK049392.1 - 3 - 4 593 84
AK082756.1 - 3 - 4 574 84
AK132204.1 - 3 - 4 571 84
BC019978.1 - 3 - 4 425 84
BC022114.1 - 3 - 4 423 84

View Full Product Details