The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164357.1 | NP_001157829.1 | 4 - 5 | 732 | 77 |
NM_001164358.1 | NP_001157830.1 | 5 - 6 | 987 | 77 | |
NM_001290558.1 | NP_001277487.1 | 4 - 5 | 601 | 77 | |
NM_146118.3 | NP_666230.2 | 5 - 6 | 768 | 77 | |
GenBank mRNA | AK049392.1 | - | 4 - 5 | 708 | 77 |
AK082756.1 | - | 4 - 5 | 689 | 77 | |
AK122568.2 | - | 5 - 6 | 952 | 77 | |
AK132204.1 | - | 4 - 5 | 686 | 77 | |
BC019978.1 | - | 4 - 5 | 540 | 77 | |
BC022114.1 | - | 4 - 5 | 538 | 77 | |
BC037109.1 | - | 5 - 6 | 728 | 77 | |
BC066998.1 | - | 5 - 6 | 955 | 77 |