The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198785.1 | NP_001185714.1 | 3 - 4 | 560 | 60 |
NM_001198787.1 | NP_001185716.1 | 3 - 4 | 332 | 60 | |
NM_007377.4 | NP_031403.2 | 3 - 4 | 843 | 60 | |
GenBank mRNA | AB093253.2 | - | 3 - 4 | 843 | 60 |
AB288871.1 | - | 3 - 4 | 462 | 60 | |
AF011908.1 | - | 3 - 4 | 299 | 60 | |
AK036705.1 | - | 3 - 4 | 334 | 60 | |
AK155100.1 | - | 3 - 4 | 334 | 60 | |
AY236858.1 | - | 3 - 4 | 338 | 60 | |
AY236859.1 | - | 3 - 4 | 365 | 60 | |
BC080846.1 | - | 3 - 4 | 414 | 60 |