The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053180.2 | NP_444410.1 | 2 - 3 | 235 | 75 |
GenBank mRNA | AY005133.1 | - | 2 - 3 | 246 | 75 |
BC031907.1 | - | 2 - 3 | 235 | 75 | |
BC058353.1 | - | - | 516 | 75 |