Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crocc
Entrez Gene ID
230872
Gene Name
ciliary rootlet coiled-coil, rootletin
Gene Aliases
-
Chromosome Location
Chr.4: 141016637 - 141060615 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145958.1 NP_001139430.1 23 - 24 3538 75
NM_172122.2 NP_742120.2 24 - 25 3791 75
GenBank mRNA AF527975.1 - 24 - 25 3791 75
AK122285.1 - 16 - 17 2478 75
BC054054.1 - 22 - 23 3519 75

View Full Product Details