Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpt1a
Entrez Gene ID
12894
Gene Name
carnitine palmitoyltransferase 1a, liver
Gene Aliases
C730027G07, CPTI, Cpt1
Chromosome Location
Chr.19: 3322326 - 3385735 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013495.2 NP_038523.2 3 - 4 374 71
GenBank mRNA AF017175.1 - - 262 71
AK050213.1 - 3 - 4 667 71
AK077454.1 - 3 - 4 328 71
AK128925.1 - 3 - 4 328 71
AK133472.1 - 3 - 4 329 71
AK136487.1 - 3 - 4 328 71
AK147770.1 - 3 - 4 71
AK149688.1 - 3 - 4 325 71
BC038395.1 - 2 - 3 309 71
BC046383.1 - 2 - 3 309 71
BC054791.1 - 3 - 4 355 71

View Full Product Details