Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dennd2a
Entrez Gene ID
209773
Gene Name
DENN/MADD domain containing 2A
Gene Aliases
B930096L08Rik, C130031G22, mKIAA1277
Chromosome Location
Chr.6: 39462374 - 39557864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172477.4 NP_766065.1 17 - 18 3268 62
GenBank mRNA AK048041.1 - 17 - 18 3253 62
AK076686.1 - 9 - 10 1413 62
AK081176.1 - 17 - 18 3269 62
AK135781.1 - 15 - 16 1813 62
AK160784.1 - 17 - 18 3030 62
AK165202.1 - 17 - 18 3123 62
AK167298.1 - 17 - 18 3122 62
BC056367.1 - 17 - 18 3138 62
BC057310.1 - 17 - 18 3138 62
BC145599.1 - 16 - 17 2947 62
BC150799.1 - 16 - 17 2947 62

View Full Product Details