The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122989.1 | NP_001116461.1 | 3 - 4 | 301 | 98 |
NM_172587.3 | NP_766175.3 | 3 - 4 | 565 | 98 | |
GenBank mRNA | AK041155.1 | - | 3 - 4 | 287 | 98 |
AK044332.1 | - | 3 - 4 | 867 | 98 | |
BC049794.1 | - | 3 - 4 | 507 | 98 | |
BC057357.1 | - | 3 - 4 | 456 | 98 | |
CN720173.1 | - | 3 - 4 | 219 | 98 |