Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc38a7
Entrez Gene ID
234595
Gene Name
solute carrier family 38, member 7
Gene Aliases
D430050E18
Chromosome Location
Chr.8: 95835920 - 95853577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172758.4 NP_766346.1 2 - 3 124 84
GenBank mRNA AK052548.1 - 2 - 3 125 84
AK171137.1 - 2 - 3 132 84
AK195271.1 - 2 - 3 122 84
AK208094.1 - 2 - 3 123 84

View Full Product Details