The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172802.4 | NP_766390.2 | 2 - 3 | 1165 | 133 |
GenBank mRNA | AK044556.1 | - | 3 - 4 | 1096 | 133 |
AK050002.1 | - | 2 - 3 | 1166 | 133 | |
BC109356.1 | - | 2 - 3 | 1126 | 133 | |
BC109357.1 | - | 2 - 3 | 1126 | 133 |