Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adamts4
Entrez Gene ID
240913
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4
Gene Aliases
3830423K05, ADAM-TS4, ADAMTS-2, ADMP-1, mKIAA0688
Chromosome Location
Chr.1: 171250422 - 171259922 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172845.2 NP_766433.1 1 - 2 1048 86
GenBank mRNA AK028364.1 - - 1048 86
AK083534.1 - 1 - 2 1048 86
AK157287.1 - - 1049 86
AK164900.1 - 1 - 2 1044 86
AK173001.1 - - 928 86
BC027773.1 - 1 - 2 682 86

View Full Product Details