Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adamts4
Entrez Gene ID
240913
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4
Gene Aliases
3830423K05, ADAM-TS4, ADAMTS-2, ADMP-1, mKIAA0688
Chromosome Location
Chr.1: 171250422 - 171259922 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172845.2 NP_766433.1 4 - 5 1673 66
GenBank mRNA AF449483.1 - 1 - 2 166 66
AK028364.1 - - 1674 66
AK076295.1 - 3 - 4 1048 66
AK083534.1 - 4 - 5 1673 66
AK156429.1 - 2 - 3 256 66
AK157287.1 - - 1674 66
AK164900.1 - 4 - 5 1669 66
AK173001.1 - - 1551 66
BC027773.1 - 4 - 5 1307 66

View Full Product Details