The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163732.1 | NP_001157204.1 | 7 - 8 | 848 | 134 |
NM_172869.4 | NP_766457.1 | 7 - 8 | 848 | 134 | |
GenBank mRNA | AK034956.1 | - | 7 - 8 | 823 | 134 |
AK048561.1 | - | 7 - 8 | 791 | 134 | |
AK079349.1 | - | 7 - 8 | 849 | 134 | |
BC037727.1 | - | 7 - 8 | 1019 | 134 | |
BC112419.1 | - | 7 - 8 | 756 | 134 | |
BC113795.1 | - | 7 - 8 | 756 | 134 | |
BC118964.2 | - | 7 - 8 | 758 | 134 |