The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173424.3 | NP_775600.1 | 3 - 4 | 1232 | 66 |
GenBank mRNA | AK078748.1 | - | 4 - 5 | 1316 | 66 |
AK084871.1 | - | 3 - 4 | 1233 | 66 | |
AK148347.1 | - | 3 - 4 | 1275 | 66 | |
BC113763.1 | - | 3 - 4 | 1211 | 66 |