Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rccd1
Entrez Gene ID
269955
Gene Name
RCC1 domain containing 1
Gene Aliases
5830436H09, E430018M08Rik
Chromosome Location
Chr.7: 80294598 - 80324696 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173445.4 NP_775621.2 8 - 9 1907 71
GenBank mRNA AK030872.1 - 8 - 9 1878 71
AK047578.1 - - 1052 71
AK084219.1 - 6 - 7 1833 71
AK088482.1 - 6 - 7 2290 71
AK088662.1 - 8 - 9 1548 71
BC069844.1 - 8 - 9 1887 71

View Full Product Details