Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam6a
Entrez Gene ID
238406
Gene Name
a disintegrin and metallopeptidase domain 6A
Gene Aliases
Adam6, tMDCIV
Chromosome Location
Chr.12: 113543908 - 113546414 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174885.3 NP_777479.2 1 - 1 153 65
GenBank mRNA AY158689.1 - 1 - 1 153 65
BC108387.1 - 1 - 1 106 65
BC139059.1 - 1 - 1 138 65

Gene Symbol
Adam6b
Entrez Gene ID
238405
Gene Name
a disintegrin and metallopeptidase domain 6B
Gene Aliases
4930523C11Rik
Chromosome Location
Chr.12: 113489565 - 113491835 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009545.1 NP_001009545.1 1 - 1 52 65
GenBank mRNA AK029742.1 - 1 - 1 107 65
AK076580.1 - 1 - 1 107 65
BC111877.1 - 1 - 1 72 65

View Full Product Details