The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168288.1 | NP_001161760.1 | 11 - 12 | 3392 | 83 |
NM_198420.2 | NP_940812.2 | 10 - 11 | 3299 | 83 | |
GenBank mRNA | AK035479.1 | - | 10 - 11 | 3097 | 83 |
AK173232.1 | - | 10 - 11 | 3234 | 83 | |
BC060065.1 | - | 6 - 7 | 1871 | 83 |