The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025290.3 | NP_079566.1 | 5 - 6 | 604 | 79 |
GenBank mRNA | AK005739.1 | - | 5 - 6 | 556 | 79 |
BC049584.1 | - | 5 - 6 | 604 | 79 | |
DQ141549.1 | - | 5 - 6 | 503 | 79 | |
DQ141550.1 | - | 5 - 6 | 503 | 79 | |
DQ141551.1 | - | 5 - 6 | 503 | 79 | |
DQ141555.1 | - | 4 - 5 | 389 | 79 |