Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dis3l2
Entrez Gene ID
208718
Gene Name
DIS3 mitotic control homolog (S. cerevisiae)-like 2
Gene Aliases
4930429A22Rik, 8030493P09Rik
Chromosome Location
Chr.1: 86703779 - 87050097 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172157.1 NP_001165628.1 20 - 21 2597 62
NM_153530.2 NP_705758.1 19 - 20 2555 62
GenBank mRNA AK029652.1 - 20 - 21 2612 62
AK031180.1 - 20 - 21 2580 62
BC036177.1 - 19 - 20 2516 62

View Full Product Details