Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhm3
Entrez Gene ID
241075
Gene Name
pleckstrin homology domain containing, family M, member 3
Gene Aliases
9430067K14Rik, A230102O09Rik, AI449570, DAPR, Plekhm1l
Chromosome Location
Chr.1: 64789120 - 64956824 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039493.1 NP_001034582.1 6 - 7 2310 99
GenBank mRNA AK034959.1 - 6 - 7 2232 99
BC063098.1 - - 2310 99

View Full Product Details