The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289614.1 | NP_001276543.1 | 1 - 2 | 220 | 92 |
NM_175938.3 | NP_787952.2 | 1 - 2 | 220 | 92 | |
GenBank mRNA | AK051061.1 | - | 1 - 2 | 206 | 92 |
AY177686.1 | - | 1 - 2 | 203 | 92 | |
BC141341.1 | - | 1 - 2 | 193 | 92 | |
CJ065534.1 | - | 1 - 2 | 220 | 92 |