The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177646.3 | NP_808314.2 | 28 - 29 | 3454 | 113 |
GenBank mRNA | AK076439.1 | - | - | 134 | 113 |
AK172901.1 | - | 24 - 25 | 2899 | 113 | |
BC048957.1 | - | 7 - 8 | 910 | 113 | |
BC057061.1 | - | 5 - 6 | 616 | 113 |