Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtmr11
Entrez Gene ID
194126
Gene Name
myotubularin related protein 11
Gene Aliases
AI035306, BC051083, Cra
Chromosome Location
Chr.3: 96161946 - 96171718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181409.3 NP_852074.2 4 - 5 563 61
GenBank mRNA AK132375.1 - 4 - 5 530 61
BC051083.1 - 4 - 5 496 61

View Full Product Details