Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudcd3
Entrez Gene ID
209586
Gene Name
NudC domain containing 3
Gene Aliases
AI427847, BC024322, mKIAA1068
Chromosome Location
Chr.11: 6105691 - 6200862 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173748.4 NP_776109.1 1 - 2 514 122
GenBank mRNA AK030050.1 - 1 - 2 239 122
AK036694.1 - 1 - 2 218 122
AK046219.1 - 1 - 2 236 122
AK049739.1 - 1 - 2 226 122
AK050476.1 - 1 - 2 204 122
AK159391.1 - 1 - 2 246 122
AK159458.1 - 1 - 2 246 122
AK169797.1 - 1 - 2 250 122
AK170329.1 - 1 - 2 247 122
AK171224.1 - 1 - 2 229 122
BC024322.1 - 1 - 2 232 122
BC037090.1 - 1 - 2 213 122
CF738060.1 - 1 - 2 478 122
EU350933.1 - 1 - 2 205 122

View Full Product Details