The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177157.4 | NP_796131.1 | 2 - 3 | 185 | 69 |
GenBank mRNA | AK028116.1 | - | 2 - 3 | 186 | 69 |
AK028122.1 | - | 2 - 3 | 186 | 69 | |
BC096406.1 | - | 2 - 3 | 173 | 69 | |
BC132450.1 | - | 2 - 3 | 181 | 69 | |
BC132478.1 | - | 2 - 3 | 181 | 69 |