Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhdc1
Entrez Gene ID
271005
Gene Name
kelch domain containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 69226917 - 69284646 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178253.5 NP_839984.1 8 - 9 790 100
GenBank mRNA AK144621.1 - 8 - 9 730 100
AY238938.1 - 8 - 9 790 100
BC119567.2 - 8 - 9 768 100
BC119568.2 - 8 - 9 774 100

View Full Product Details