Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a6
Entrez Gene ID
330836
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 6
Gene Aliases
AI643885, LAT-2, LAT3
Chromosome Location
Chr.8: 106168830 - 106198708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178798.3 NP_848913.1 7 - 8 1236 61
GenBank mRNA AK030762.1 - 8 - 9 1038 61
AK046548.1 - 6 - 7 1903 61
AK049478.1 - 7 - 8 1236 61
AK053899.1 - 7 - 8 1245 61
AK088592.1 - 7 - 8 1153 61
AK129099.1 - 4 - 5 2632 61
AK157127.1 - 7 - 8 1375 61
AK157379.1 - 6 - 7 1905 61
AK164666.1 - 7 - 8 1237 61
AK165629.1 - 7 - 8 1253 61
AK165979.1 - 7 - 8 1255 61
AK191440.1 - 3 - 4 263 61
BC038404.1 - 6 - 7 1171 61
BC042898.1 - 4 - 5 1768 61

View Full Product Details