Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid5a
Entrez Gene ID
214855
Gene Name
AT rich interactive domain 5A (MRF1-like)
Gene Aliases
D430024K22Rik, Mrf1
Chromosome Location
Chr.1: 36307706 - 36324036 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172205.1 NP_001165676.1 1 - 2 109 80
NM_145996.4 NP_666108.2 1 - 2 109 80
NR_033310.1 1 - 2 109 80
GenBank mRNA AK029237.1 - 1 - 2 88 80
AK085015.1 - 1 - 2 89 80
AK149839.1 - 1 - 2 89 80
AK155555.1 - 1 - 2 86 80
AK156376.1 - 1 - 2 89 80
AK156934.1 - 1 - 2 89 80
AK157607.1 - 1 - 2 89 80
BC027152.1 - 1 - 2 73 80

View Full Product Details