Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4b1
Entrez Gene ID
69810
Gene Name
C-type lectin domain family 4, member b1
Gene Aliases
1810046I24Rik, Aplra2, Clec4b, DCAR, DCARbeta, Dcar2
Chromosome Location
Chr.6: 123049962 - 123071555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027218.3 NP_081494.1 2 - 3 251 89
GenBank mRNA AA914236.1 - 2 - 3 251 89
AK007794.1 - 2 - 3 208 89
AY230260.1 - 2 - 3 215 89

View Full Product Details