The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013775.1 | NP_038803.1 | 1 - 2 | 198 | 140 |
GenBank mRNA | AF195489.1 | - | 1 - 2 | 198 | 140 |
AK139548.1 | - | 1 - 2 | 213 | 140 | |
BC099461.1 | - | 1 - 2 | 182 | 140 |