Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prpf38b
Entrez Gene ID
66921
Gene Name
PRP38 pre-mRNA processing factor 38 (yeast) domain containing B
Gene Aliases
1110021E09Rik, AU018955
Chromosome Location
Chr.3: 108902807 - 108911752 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025845.2 NP_080121.1 4 - 5 817 67
GenBank mRNA AK015938.1 - 4 - 5 824 67
AK018126.1 - 4 - 5 841 67
AK159267.1 - 4 - 5 840 67
AK181054.1 - 3 - 4 224 67
BC050900.1 - 4 - 5 817 67
BC052077.1 - 4 - 5 817 67

View Full Product Details