The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111304.1 | NP_001104774.1 | 20 - 21 | 3803 | 151 |
NM_027758.4 | NP_082034.1 | 17 - 18 | 3104 | 151 | |
GenBank mRNA | AK017015.1 | - | 6 - 7 | 622 | 151 |
AK049049.1 | - | 17 - 18 | 3105 | 151 | |
AK077081.1 | - | 6 - 7 | 636 | 151 | |
AK134614.1 | - | 20 - 21 | 3080 | 151 | |
AK173050.1 | - | 20 - 21 | 2976 | 151 | |
BC057027.1 | - | 16 - 17 | 2391 | 151 |