Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfyc
Entrez Gene ID
18046
Gene Name
nuclear transcription factor-Y gamma
Gene Aliases
-
Chromosome Location
Chr.4: 120757435 - 120831579 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048168.2 NP_001041633.1 5 - 6 529 143
NM_008692.4 NP_032718.2 5 - 6 602 143
GenBank mRNA AB006181.1 - 4 - 5 382 143
BC020117.1 - 5 - 6 528 143
BC053723.1 - 5 - 6 532 143
BC085261.1 - 5 - 6 528 143
BM461330.1 - 3 - 4 233 143

View Full Product Details