The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134123.3 | NP_598884.2 | 16 - 17 | 2340 | 69 |
GenBank mRNA | AK032418.1 | - | - | 542 | 69 |
AK122451.1 | - | 16 - 17 | 2207 | 69 | |
AK161113.1 | - | 2 - 3 | 252 | 69 | |
BC030846.1 | - | 3 - 4 | 338 | 69 | |
BC038363.1 | - | 16 - 17 | 1984 | 69 | |
BC075621.1 | - | 16 - 17 | 2333 | 69 |