The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170691.1 | NP_001164162.1 | 5 - 6 | 535 | 75 |
NM_178736.5 | NP_848851.3 | 5 - 6 | 540 | 75 | |
GenBank mRNA | AK029044.1 | - | 5 - 6 | 511 | 75 |
AK036607.1 | - | 5 - 6 | 518 | 75 | |
AK036746.1 | - | 5 - 6 | 480 | 75 | |
AK041137.1 | - | 5 - 6 | 441 | 75 | |
AK082730.1 | - | 4 - 5 | 307 | 75 | |
AK084950.1 | - | 5 - 6 | 540 | 75 | |
BC079654.1 | - | 5 - 6 | 501 | 75 |