Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gclc
Entrez Gene ID
14629
Gene Name
glutamate-cysteine ligase, catalytic subunit
Gene Aliases
D9Wsu168e, GLCL-H, Ggcs-hs, Glclc
Chromosome Location
Chr.9: 77754516 - 77794489 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010295.2 NP_034425.1 6 - 7 1032 87
GenBank mRNA AK144339.1 - 6 - 7 1033 87
AK160876.1 - 6 - 7 792 87
AK169676.1 - 6 - 7 1005 87
BC019374.1 - 6 - 7 789 87
U85414.1 - 6 - 7 757 87
U85498.1 - 6 - 7 757 87

View Full Product Details