The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023672.2 | NP_076161.2 | 15 - 16 | 1396 | 93 |
NM_198438.1 | NP_940840.1 | 14 - 15 | 1315 | 93 | |
GenBank mRNA | AF500117.1 | - | 14 - 15 | 1045 | 93 |
AK011509.1 | - | 15 - 16 | 1011 | 93 | |
AK011853.1 | - | 15 - 16 | 1366 | 93 | |
AK017711.1 | - | 15 - 16 | 1400 | 93 | |
AK041388.1 | - | 11 - 12 | 912 | 93 | |
AK153733.1 | - | 11 - 12 | 756 | 93 | |
AY167987.1 | - | 14 - 15 | 928 | 93 | |
BC003430.1 | - | 14 - 15 | 1069 | 93 | |
BC035475.1 | - | 14 - 15 | 1258 | 93 |