The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183289.3 | NP_899112.2 | 3 - 4 | 735 | 66 |
GenBank mRNA | AK017700.1 | - | 3 - 4 | 735 | 66 |
BC107234.1 | - | 3 - 4 | 350 | 66 | |
BC107235.1 | - | 3 - 4 | 350 | 66 |