Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
19175
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Lmp19, Mpnd
Chromosome Location
Chr.11: 70525357 - 70527858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008946.4 NP_032972.3 4 - 5 464 181
GenBank mRNA AK078437.1 - 4 - 5 436 181
AK132042.1 - 4 - 5 447 181
AK167123.1 - 4 - 5 436 181
AK167227.1 - 4 - 5 432 181
BC013897.1 - 4 - 5 424 181
U13393.1 - 4 - 5 385 181

View Full Product Details