Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fdps
Entrez Gene ID
110196
Gene Name
farnesyl diphosphate synthetase
Gene Aliases
6030492I17Rik, AI256750, Fdpsl1, mKIAA1293
Chromosome Location
Chr.3: 89093588 - 89101967 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253751.1 NP_001240680.1 4 - 5 628 175
NM_134469.4 NP_608219.1 3 - 4 412 175
GenBank mRNA AF309508.1 - 2 - 3 317 175
AK088601.1 - 3 - 4 332 175
AK140881.1 - 3 - 4 404 175
AK151742.1 - 3 - 4 341 175
AK166026.1 - 3 - 4 331 175
AK168008.1 - 3 - 4 331 175
BC048497.1 - 3 - 4 390 175
BC087886.1 - 3 - 4 335 175
CT010365.1 - 2 - 3 284 175

View Full Product Details